Methyl-2-chloropyrimidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

074038-1G 074038-5G
102534-306EA 75.38 USD
102534-306 102534-308
Methyl-2-chloropyrimidine-4-carboxylate ≥95%
Methyl-2-chloropyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD11841080
CAS Number: 149849-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR