1-N-Boc-pyrrolidin-2-ylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-N-Boc-Pyrrolidin-2-ylboronic acid, 2-Borono-1-pyrrolidinecarboxylic acid 1-(1,1-dimethylethyl) ester

074037-1G 074037-5G
102534-300EA 61.54 USD
102534-300 102534-302
1-N-Boc-pyrrolidin-2-ylboronic acid ≥95%
1-N-Boc-pyrrolidin-2-ylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₈BNO₄
MW: 215.0545 g/mol
MDL Number: MFCD02183523
CAS Number: 149682-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR