1-Boc-2-piperidineacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-2-Piperidineacetic acid

074035-1G 074035-5G
102534-290EA 320 USD
102534-290 102534-292
1-Boc-2-piperidineacetic acid ≥95%
1-Boc-2-piperidineacetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02179000
CAS Number: 149518-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR