5-Bromo-2-chloro-4-fluorophenol ≥95%

Supplier: Matrix Scientific

074034-1G 074034-5G
102534-286EA 153.85 USD
102534-286 102534-288
5-Bromo-2-chloro-4-fluorophenol ≥95%
5-Bromo-2-chloro-4-fluorophenol

Hazard: Irritant

Formula: C₆H₃BrClFO
MW: 225.4428 g/mol
MDL Number: MFCD08436979
CAS Number: 148254-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR