5-Bromo-1,3-difluoro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Difluoro-4-nitrobromobenzene

074033-1G 074033-5G
102534-278EA 23.08 USD
102534-278 102534-280
5-Bromo-1,3-difluoro-2-nitrobenzene ≥95%
5-Bromo-1,3-difluoro-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₂BrF₂NO₂
MW: 237.9864 g/mol
MDL Number: MFCD12025434
CAS Number: 147808-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR