2-Phenyldihydro-2H-pyran-4(3H)-one ≥95%

Supplier: Matrix Scientific

074032-250MG 074032-1G 074032-5G
102534-272EA 400 USD
102534-272 102534-274 102534-276
2-Phenyldihydro-2H-pyran-4(3H)-one ≥95%
2-Phenyldihydro-2H-pyran-4(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.2118 g/mol
MDL Number: MFCD11112124
CAS Number: 147688-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR