Tenofovir ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid, (R)-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid

074030-1G 074030-5G 074030-25G
102534-262EA 58.46 USD
102534-262 102534-264 102551-860
Tenofovir ≥95%
Tenofovir

Hazard: Irritant

Formula: C₉H₁₄N₅O₄P
MW: 287.22 g/mol
MDL Number: MFCD00943794
CAS Number: 147127-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR