2-Bromo-5-chlorotoluene ≥95%
Supplier: Matrix Scientific
074027-10G
074027-100G
102534-246EA
55.38
USD
102534-246
102534-248
2-Bromo-5-chlorotoluene ≥95%
2-Bromo-5-chlorotoluene
Hazard: Irritant
Formula:
C₇H₆BrCl |
MDL Number:
MFCD00000602 CAS Number: 14495-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...