5-(2-Bromophenyl)-1-(triphenylmethyl)-1H-tetrazole ≥95%

Supplier: Matrix Scientific

074024-1G 074024-5G 074024-10G
102534-232EA 178.46 USD
102534-232 102534-234 102534-236
5-(2-Bromophenyl)-1-(triphenylmethyl)-1H-tetrazole ≥95%
5-(2-Bromophenyl)-1-(triphenylmethyl)-1H-tetrazole

Hazard: Irritant

Formula: C₂₆H₁₉BrN₄
CAS Number: 143945-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR