2,6-Diaminopyridine ≥95%

Supplier: Matrix Scientific

074023-10G 074023-100G
102534-228EA 36.92 USD
102534-228 102534-230
2,6-Diaminopyridine ≥95%
2,6-Diaminopyridine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD00006329
CAS Number: 141-86-6
UN: 2671
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR