tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(((trifluoromethyl)sulphonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate, 1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridin-4-yl trifluoromethanesulphonate

074020-1G 074020-5G
102534-214EA 153.85 USD
102534-214 102534-216
tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate ≥95%
tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₆F₃NO₅S
MW: 331.3087 g/mol
MDL Number: MFCD09997858
CAS Number: 138647-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR