Piperidin-3-amine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

074018-1G 074018-5G
102534-206EA 233.85 USD
102534-206 102534-208
Piperidin-3-amine dihydrochloride ≥95%
Piperidin-3-amine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₄Cl₂N₂
MW: 173.081 g/mol
MDL Number: MFCD00012773
CAS Number: 138060-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR