2-Acetyl-5-methylthiophene ≥95%

Supplier: Matrix Scientific

074017-1G 074017-5G 074017-10G
102534-200EA 55.38 USD
102534-200 102534-202 102534-204
2-Acetyl-5-methylthiophene ≥95%
2-Acetyl-5-methylthiophene

Hazard: Irritant

Formula: C₇H₈OS
MW: 140.21 g/mol
MDL Number: MFCD00014529
CAS Number: 13679-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR