(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-N-Boc-2-hydroxymethylmorpholine, (R)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate

074015-1G 074015-5G
102534-190EA 60 USD
102534-190 102534-192
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate ≥95%
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
MDL Number: MFCD09260605
CAS Number: 135065-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR