2,4-Dimethoxyphenol ≥95%

Supplier: Matrix Scientific

074013-1G 074013-5G 074013-10G
102534-178EA 153.85 USD
102534-178 102534-180 102534-182
2,4-Dimethoxyphenol ≥95%
2,4-Dimethoxyphenol

Hazard: Irritant

Formula: C₈H₁₀O₃
MW: 154.1632 g/mol
MDL Number: MFCD00060637
CAS Number: 13330-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR