Imidazo[1,2-a]pyridin-6-ylmethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(Hydroxymethyl)imidazo[1,2-a]pyridine
074011-1G
074011-5G
074011-10G
102534-166EA
303.08
USD
102534-166
102534-168
102534-170
Imidazo[1,2-a]pyridin-6-ylmethanol ≥95%
Imidazo[1,2-a]pyridin-6-ylmethanol
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.1600 g/mol |
MDL Number:
MFCD07368521 CAS Number: 132213-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...