2-Bromo-3-formylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde
074006-5G
074006-25G
102534-142EA
52.31
USD
102534-142
102833-666
2-Bromo-3-formylpyridine ≥95%
2-Bromo-3-formylpyridine
Hazard: Irritant/Refrigerate
Formula:
C₆H₄BrNO MW: 186.01 g/mol |
MDL Number:
MFCD04966945 CAS Number: 128071-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...