2-Amino-3,5-difluorobenzoic acid ≥95%

Supplier: Matrix Scientific

074004-1G 074004-5G
102534-130EA 92.31 USD
102534-130 102534-132
2-Amino-3,5-difluorobenzoic acid ≥95%
2-Amino-3,5-difluorobenzoic acid

Hazard: Irritant

Formula: C₇H₅F₂NO₂
MDL Number: MFCD01569541
CAS Number: 126674-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR