1-Benzylazepan-4-one ≥95%

Supplier: Matrix Scientific

073996-1G 073996-5G
102534-096EA 461.54 USD
102534-096 102534-098
1-Benzylazepan-4-one ≥95%
1-Benzylazepan-4-one

Hazard: Irritant

Formula: C₁₃H₁₇NO
MDL Number: MFCD04038554
CAS Number: 1208-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR