Methylpyrazin-2-ylmethylamine ≥95%

Supplier: Matrix Scientific

073995-1G 073995-5G
102534-092EA 710.77 USD
102534-092 102534-094
Methylpyrazin-2-ylmethylamine ≥95%
Methylpyrazin-2-ylmethylamine

Hazard: Irritant

Formula: C₆H₉N₃
MDL Number: MFCD06738965
CAS Number: 120739-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR