4-Chloro-3,5-dinitrobenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dinitro-4-chlorobenzoic acid

073992-10G 073992-100G
102534-078EA 23.08 USD
102534-078 102534-080
4-Chloro-3,5-dinitrobenzoic acid ≥95%
4-Chloro-3,5-dinitrobenzoic acid

Hazard: Irritant

Formula: C₇H₃ClN₂O₆
MW: 246.56 g/mol
Boiling Pt: 315°C
Melting Pt: 159-162°C
Flash Pt: 186°C
MDL Number: MFCD00007080
CAS Number: 118-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR