(R)-3-Aminotetrahydrofuran hydrochloride ≥95%

Supplier: Matrix Scientific

073991-1G 073991-5G
102534-074EA 147.69 USD
102534-074 102534-076
(R)-3-Aminotetrahydrofuran hydrochloride ≥95%
(R)-3-Aminotetrahydrofuran hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MW: 123.5800 g/mol
MDL Number: MFCD08461720
CAS Number: 1072015-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR