3-Bromo-4-fluoropyridine ≥95%

Supplier: Matrix Scientific

073990-1G 073990-5G
102534-068EA 121.54 USD
102534-068 102534-070
3-Bromo-4-fluoropyridine ≥95%
3-Bromo-4-fluoropyridine

Hazard: Irritant/Store in Freezer

Formula: C₅H₃BrFN
MDL Number: MFCD04114101
CAS Number: 116922-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR