3-Bromo-5-(trifluoromethyl)phenylacetic acid ≥95%

Supplier: Matrix Scientific

073988-1G 073988-5G
102534-056EA 116.92 USD
102534-056 102534-058
3-Bromo-5-(trifluoromethyl)phenylacetic acid ≥95%
3-Bromo-5-(trifluoromethyl)phenylacetic acid

Hazard: Irritant

Formula: C9H6BrF3O2
MW: 283.04 g/mol
MDL Number: MFCD13194218
CAS Number: 1161362-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR