(S)-2-Hydroxy-4-phenylbutyric acid ≥95%

Supplier: Matrix Scientific

073985-1G 073985-5G 073985-10G
102534-042EA 284.62 USD
102534-042 102534-044 102534-046
(S)-2-Hydroxy-4-phenylbutyric acid ≥95%
(S)-2-Hydroxy-4-phenylbutyric acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.2005 g/mol
MDL Number: MFCD06656492
CAS Number: 115016-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR