5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

073984-250MG 073984-1G 073984-5G
102534-036EA 498.46 USD
102534-036 102534-038 102534-040
5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClN
MDL Number: MFCD09026771
CAS Number: 1073968-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR