3,5-Dimethoxybenzoic acid ≥95%
Supplier: Matrix Scientific
073981-10G
073981-100G
102534-022EA
27.69
USD
102534-022
102534-024
3,5-Dimethoxybenzoic acid ≥95%
3,5-Dimethoxybenzoic acid
Hazard: Irritant
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol |
MDL Number:
MFCD00002502 CAS Number: 1132-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...