2,4-Difluoro-m-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Difluoro-3-methylbenzoic acid

073980-1G 073980-5G
102534-018EA 569.23 USD
102534-018 102534-020
2,4-Difluoro-m-toluic acid ≥95%
2,4-Difluoro-m-toluic acid

Hazard: Irritant

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD03265223
CAS Number: 112857-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR