3,4-Dibromo-1H-pyrrole-2,5-dione ≥95%
Supplier: Matrix Scientific
Synonyms:
3,4-Dibromomaleimide
073978-1G
073978-5G
073978-10G
102534-006EA
90.77
USD
102534-006
102534-008
102534-010
3,4-Dibromo-1H-pyrrole-2,5-dione ≥95%
3,4-Dibromo-1H-pyrrole-2,5-dione
Hazard: Irritant
Formula:
C4HBr2NO2 MW: 254.8600 g/mol |
MDL Number:
MFCD00185696 CAS Number: 1122-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...