3-Amino-2-chlorobenzoic acid ≥95%

Supplier: Matrix Scientific

073974-1G 073974-5G 073974-10G
102533-982EA 355.38 USD
102533-982 102533-984 102533-986
3-Amino-2-chlorobenzoic acid ≥95%
3-Amino-2-chlorobenzoic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.5810 g/mol
MDL Number: MFCD00191726
CAS Number: 108679-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR