(1-Aminocyclopropyl)methanol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Amino-1-(hydroxymethyl)cyclopropane hydrochloride, (1-Amino-cyclopropyl)-methanol hydrochloride

073972-250MG 073972-1G 073972-5G
102533-970EA 364.62 USD
102533-970 102533-972 102533-974
(1-Aminocyclopropyl)methanol hydrochloride ≥95%
(1-Aminocyclopropyl)methanol hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MW: 123.5813 g/mol
MDL Number: MFCD09834330
CAS Number: 115652-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR