4-(4-Hydroxyphenyl)cyclohexanone ≥95%

Supplier: Matrix Scientific

073970-10G 073970-100G
102533-960EA 249.23 USD
102533-960 102533-962
4-(4-Hydroxyphenyl)cyclohexanone ≥95%
4-(4-Hydroxyphenyl)cyclohexanone

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00210693
CAS Number: 105640-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR