(R)-3-Hydroxypyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Pyrrolidinol hydrochloride, (3R)-pyrrolidin-3-ol hydrochloride, (R)-3-Hydroxypyrrolidine HCl

073969-5G 073969-10G 073969-100G
102533-954EA 143.08 USD
102533-954 102533-956 102533-958
(R)-3-Hydroxypyrrolidine hydrochloride ≥95%
(R)-3-Hydroxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD00191570
CAS Number: 104706-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR