2,5-Difluoro-p-toluic acid ≥95%
Supplier: Matrix Scientific
073967-1G
073967-5G
102533-948EA
30.77
USD
102533-948
102533-950
2,5-Difluoro-p-toluic acid ≥95%
2,5-Difluoro-p-toluic acid
Hazard: Irritant
Formula:
C₈H₆F₂O₂ |
MDL Number:
MFCD08063230 CAS Number: 103877-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...