2,4,6-Triaminopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrimidine-2,4,6-triyltriamine

073961-10G 073961-100G
102533-918EA 47.69 USD
102533-918 102533-920
2,4,6-Triaminopyrimidine ≥95%
2,4,6-Triaminopyrimidine

Hazard: Irritant

Formula: C₄H₇N₅
MW: 125.13 g/mol
MDL Number: MFCD00006100
CAS Number: 1004-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR