1-Bromo-2,3-difluoro-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Difluoro-4-bromonitrobenzene
073960-250MG
073960-1G
073960-5G
102533-912EA
340
USD
102533-912
102533-914
102533-916
1-Bromo-2,3-difluoro-4-nitrobenzene ≥95%
1-Bromo-2,3-difluoro-4-nitrobenzene
Hazard: Irritant
Formula:
C₆H₂BrF₂NO₂ MW: 237.9900 g/mol |
MDL Number:
MFCD09839215 CAS Number: 1003708-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...