2-Iodo-6-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

073959-1G 073959-5G
102533-908EA 198.46 USD
102533-908 102533-910
2-Iodo-6-(trifluoromethyl)pyridine ≥95%
2-Iodo-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃F₃IN
CAS Number: 100366-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR