3-Fluoro-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

073958-1G 073958-5G
102533-902EA 44.62 USD
102533-902 102533-904
3-Fluoro-2-nitrobenzoic acid ≥95%
3-Fluoro-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD07368342
CAS Number: 1000339-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR