2-Amino-5-(trifluoromethyl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-cyanobenzotrifluoride, 2-Amino-5-trifluoromethylbenzonitrile

073957-5G 073957-25G
102533-898EA 135.38 USD
102533-898 102663-964
2-Amino-5-(trifluoromethyl)benzonitrile ≥95%
2-Amino-5-(trifluoromethyl)benzonitrile

Hazard: Toxic

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Boiling Pt: 95-115°C/0.4-0.7 Torr
Melting Pt: 72-74°C
MDL Number: MFCD03407465
CAS Number: 6526-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR