N-Methyl-2-thiazolamine ≥95%

Supplier: Matrix Scientific

073956-250MG 073956-1G 073956-5G
102533-890EA 364.62 USD
102533-890 102533-892 102533-894
N-Methyl-2-thiazolamine ≥95%
N-Methyl-2-thiazolamine

Hazard: Irritant

Formula: C₄H₆N₂S
MW: 114.1700 g/mol
MDL Number: MFCD09966061
CAS Number: 6142-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR