10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine ≥95%

Supplier: Matrix Scientific

Synonyms: Quetiapine Related Compound G, Dibenzo[b,f][1,4]thiazepin-11(10H)-one

073954-5G 073954-10G 073954-100G
102533-878EA 72.31 USD
102533-878 102533-880 102533-882
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine ≥95%
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine

Hazard: Irritant

Formula: C₁₃H₉NOS
MW: 227.2817 g/mol
Melting Pt: 257…259 °C
MDL Number: MFCD00901197
CAS Number: 3159-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR