1-(3,4-Dichlorophenyl)urea ≥95%
Supplier: Matrix Scientific
073952-1G
073952-5G
102533-868EA
230.77
USD
102533-868
102533-870
1-(3,4-Dichlorophenyl)urea ≥95%
1-(3,4-Dichlorophenyl)urea
Hazard: Irritant
Formula:
C₇H₆Cl₂N₂O Melting Pt: 155…156 °C |
MDL Number:
MFCD00025417 CAS Number: 2327-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...