3-Aminopiperidin-2-one ≥95%

Supplier: Matrix Scientific

073949-1G 073949-5G
102533-854EA 135.38 USD
102533-854 102533-856
3-Aminopiperidin-2-one ≥95%
3-Aminopiperidin-2-one

Hazard: Irritant

Formula: C5H10N2O
MDL Number: MFCD06809610
CAS Number: 1892-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR