Dihydro-2H-pyran-3(4H)-one ≥95%

Supplier: Matrix Scientific

073946-1G 073946-5G
102533-840EA 569.23 USD
102533-840 102533-842
Dihydro-2H-pyran-3(4H)-one ≥95%
Dihydro-2H-pyran-3(4H)-one

Hazard: Irritant

Formula: C₅H₈O₂
MW: 100.1158 g/mol
MDL Number: MFCD00182426
CAS Number: 23462-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR