4-Bromo-3-methoxypyridine ≥95%

Supplier: Matrix Scientific

073945-250MG 073945-1G
102533-834EA 330.85 USD
102533-834 102533-836
4-Bromo-3-methoxypyridine ≥95%
4-Bromo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₆BrNO
MDL Number: MFCD08234926
CAS Number: 109911-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR