4,6-Dichloropicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4,6-Dichloropyridine-2-carboxylic acid

073944-1G 073944-5G
102533-830EA 130.77 USD
102533-830 102533-832
4,6-Dichloropicolinic acid ≥95%
4,6-Dichloropicolinic acid

Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192.0000 g/mol
MDL Number: MFCD09910267
CAS Number: 88912-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR