2,4,6-Trichloroquinazoline ≥95%

Supplier: Matrix Scientific

073943-250MG 073943-1G 073943-5G
102533-824EA 340 USD
102533-824 102533-826 102533-828
2,4,6-Trichloroquinazoline ≥95%
2,4,6-Trichloroquinazoline

Hazard: Toxic

Formula: C₈H₃Cl₃N₂
MW: 233.4818 g/mol
MDL Number: MFCD08445622
CAS Number: 20028-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR