Methyl-4-acetamido-5-chloro-2-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-acetamido-5-chloro-2-hydroxybenzoate

073939-1G 073939-5G 073939-10G
102533-806EA 355.38 USD
102533-806 102533-808 102533-810
Methyl-4-acetamido-5-chloro-2-hydroxybenzoate ≥95%
Methyl-4-acetamido-5-chloro-2-hydroxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₄
MW: 243.6400 g/mol
MDL Number: MFCD02093078
CAS Number: 24190-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR