2-Methyl-3-nitroanisole ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methoxy-2-methyl-3-nitrobenzene, Methyl 3-nitro-o-tolyl ether, 2-Methoxy-6-nitrotoluene
073938-5G
073938-25G
102533-802EA
110.77
USD
102533-802
102833-662
2-Methyl-3-nitroanisole ≥95%
2-Methyl-3-nitroanisole
Hazard: Irritant
Formula:
C₈H₉NO₃ MW: 167.16 g/mol |
MDL Number:
MFCD00007159 CAS Number: 4837-88-1 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...