4-Amino-3,5-dinitropyridine ≥95%

Supplier: Matrix Scientific

073937-1G 073937-5G 073937-10G
102533-794EA 72.31 USD
102533-794 102533-796 102533-798
4-Amino-3,5-dinitropyridine ≥95%
4-Amino-3,5-dinitropyridine

Hazard: Irritant

Formula: C₅H₄N₄O₄
MW: 184.1097 g/mol
MDL Number: MFCD00160302
CAS Number: 31793-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR